Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps6kb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031985.1 | NP_114191.1 | 3 | 273 | ||
XM_006247111.2 | XP_006247173.1 | 3 | 410 | |||
XM_008768120.1 | XP_008766342.1 | 3 | 674 | |||
XM_008768121.1 | XP_008766343.1 | 4 | 296 | |||
GenBank mRNA | BC089789.1 | - | 3 | 300 | ||
M58340.1 | - | 3 | 273 |