Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCNC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013399.1 | NP_001013417.1 | 2 | 175 | ||
NM_005190.3 | NP_005181.2 | 2 | 397 | |||
XM_006715594.1 | XP_006715657.1 | 2 | 556 | |||
XM_011536232.2 | XP_011534534.1 | 2 | 556 | |||
XM_017011436.1 | XP_016866925.1 | 2 | 556 | |||
GenBank mRNA | AB125137.1 | - | 2 | 184 | ||
AK222768.1 | - | 2 | 211 | |||
AK223247.1 | - | 2 | 198 | |||
AK291914.1 | - | 2 | 206 | |||
BC010135.2 | - | 2 | 230 | 4106819 | ||
BC026272.1 | - | 2 | 211 | |||
BC041123.1 | - | 2 | 380 | 5759974 | ||
BC056153.1 | - | 2 | 219 | 6157106 | ||
BM788717.1 | - | 2 | 175 | |||
M74091.1 | - | 2 | 198 |