Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmga2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032070.1 | NP_114459.1 | 3, 4 | 662 | ||
GenBank mRNA | U89695.1 | - | 3, 4 | 662 |