Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034164.1 | NP_001029336.1 | 4, 5 | 571 | ||
NM_032612.3 | NP_116001.2 | 4, 5 | 571 | |||
GenBank mRNA | AF205604.1 | - | 2, 3 | 281 | ||
BC062079.1 | - | 4, 5 | 571 | 5599645 | ||
CV075250.1 | - | 4, 5 | 489 |