Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab11b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032617.2 | NP_116006.1 | 4 | 544 | ||
GenBank mRNA | AF286534.1 | - | 4 | 550 | ||
BC062041.1 | - | 4 | 544 | 5598168 | ||
D01046.1 | - | 4 | 530 |