Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mdh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033235.1 | NP_150238.1 | 4 | 339 | ||
GenBank mRNA | AF093773.1 | - | 4 | 339 | ||
BC059124.1 | - | 4 | 380 | 6921518 | ||
FQ212702.1 | - | 4 | 383 | |||
FQ215129.1 | - | 4 | 393 | |||
FQ215603.1 | - | 4 | 378 | |||
FQ217383.1 | - | 4 | 383 | |||
FQ217819.1 | - | 383 | ||||
FQ224891.1 | - | 4 | 381 | |||
FQ233582.1 | - | 4 | 383 |