Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033376.1 | NP_203694.1 | 2 | 432 | ||
GenBank mRNA | AB048823.1 | - | 3 | 285 | ||
AB048824.1 | - | 3 | 274 | |||
AF031384.1 | - | 2 | 432 |