Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sult1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031834.1 | NP_114022.1 | 7 | 678 | ||
GenBank mRNA | AF394783.1 | - | 6 | 601 | ||
FQ209630.1 | - | 7 | 687 | |||
FQ210886.1 | - | 7 | 683 | |||
FQ214244.1 | - | 7 | 683 | |||
FQ218658.1 | - | 7 | 682 | |||
FQ219178.1 | - | 7 | 683 | |||
FQ219521.1 | - | 7 | 683 | |||
FQ219542.1 | - | 7 | 683 | |||
L19998.1 | - | 7 | 678 | |||
X52883.1 | - | 7 | 666 | |||
X68640.1 | - | 6 | 583 |