Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndrg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271091.1 | NP_001258020.1 | 15 | 1103 | ||
NM_001271092.1 | NP_001258021.1 | 15 | 1103 | |||
NM_001271093.1 | NP_001258022.1 | 15 | 1050 | |||
NM_001271094.1 | NP_001258023.1 | 15 | 1050 | |||
NM_001271095.1 | NP_001258024.1 | 13 | 968 | |||
NM_031967.2 | NP_114173.2 | 13 | 968 | |||
XM_006255123.2 | XP_006255185.1 | 15 | 1045 | |||
GenBank mRNA | AF045564.1 | - | 13 | 900 | ||
AY217029.1 | - | 13 | 939 | |||
AY217030.1 | - | 15 | 1050 | |||
AY217031.1 | - | 15 | 1050 | |||
AY217032.1 | - | 15 | 1098 | |||
AY217033.1 | - | 15 | 1098 | |||
AY255791.1 | - | 13 | 939 | |||
CB585874.1 | - | 9 | 525 | |||
CB713901.1 | - | 6 | 283 |