Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trpv1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031982.1 | NP_114188.1 | 9, 10 | 1562 | ||
XM_008767882.1 | XP_008766104.1 | 8, 9 | 1569 | |||
GenBank mRNA | AB015231.3 | - | 813 | |||
AB040873.1 | - | 8, 9 | 1482 | |||
AB041029.1 | - | 7, 8 | 1302 | |||
AF029310.1 | - | 9, 10 | 1562 | |||
AF158248.2 | - | 3, 4 | 895 | |||
LC008303.1 | - | 4, 5 | 802 |