Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TLN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006289.3 | NP_006280.3 | 5 | 776 | ||
GenBank mRNA | AB028950.2 | - | 5 | 751 | ||
AF177198.1 | - | 5 | 512 | |||
BC042923.1 | - | 5 | 562 | 5576203 |