Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cxcr5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053303.1 | NP_445755.1 | 2 | 113 | ||
XM_006242901.2 | XP_006242963.1 | 3 | 736 |