Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Camlg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053334.1 | NP_445786.1 | 1 | 172 | ||
XM_006253658.2 | XP_006253720.1 | 1 | 211 | |||
XM_006253659.2 | XP_006253721.1 | 1 | 212 | |||
XM_006253660.2 | XP_006253722.1 | 1 | 215 | |||
XM_008771508.1 | XP_008769730.1 | 1 | 215 | |||
XM_008771509.1 | XP_008769731.1 | 1 | 213 | |||
XM_008771510.1 | XP_008769732.1 | 1 | 213 | |||
GenBank mRNA | AF302085.1 | - | 1 | 172 | ||
BC076377.1 | - | 1 | 171 | 7110159 |