Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cacng2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053351.1 | NP_445803.1 | 1 | 206 | ||
GenBank mRNA | AF118818.1 | - | 1 | 206 | ||
AF361339.1 | - | 1 | 173 |