Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Timm8a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053370.3 | NP_445822.1 | 214 | |||
GenBank mRNA | AF150082.1 | - | 215 | |||
BC060552.1 | - | 219 | 6921445 |