Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slpi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053372.1 | NP_445824.1 | 1, 2 | 93 | ||
GenBank mRNA | AF151982.1 | - | 1, 2 | 93 | ||
AF178426.1 | - | 1, 2 | 89 | |||
AF421377.1 | - | 1, 2 | 103 |