Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CASP14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012114.2 | NP_036246.1 | 4 | 406 | ||
XM_011527861.1 | XP_011526163.1 | 4 | 406 | |||
GenBank mRNA | AF097874.1 | - | 3 | 242 | ||
AK313992.1 | - | 4 | 406 | |||
BC069541.1 | - | 3 | 242 | 7262238 | ||
BC103868.1 | - | 3 | 242 | 40003517 | ||
BC103869.1 | - | 3 | 242 | 40003519 | ||
EU262664.1 | - | 3 | 242 | |||
EU502748.1 | - | 4 | 502 |