Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dclk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053343.3 | NP_445795.1 | 4, 5 | 448 | ||
XM_008761004.1 | XP_008759226.1 | 4, 5 | 2350 | |||
XM_008761005.1 | XP_008759227.1 | 4, 5 | 1376 | |||
XM_008761006.1 | XP_008759228.1 | 4, 5 | 2353 | |||
XM_008761007.1 | XP_008759229.1 | 4, 5 | 460 | |||
GenBank mRNA | FQ211799.1 | - | 4, 5 | 435 | ||
U78857.1 | - | 4, 5 | 438 |