Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col1a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053356.1 | NP_445808.1 | 360 | |||
GenBank mRNA | AF121217.1 | - | 360 | |||
BC108298.1 | - | 454 |
Gene Symbol: NEWGENE_621351 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008762736.1 | XP_008760958.1 | 8 | 710 |