Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ssbp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053358.1 | NP_445810.1 | 5, 6 | 708 | ||
XM_006238536.2 | XP_006238598.1 | 5, 6 | 741 | |||
XM_006238537.2 | XP_006238599.1 | 5, 6 | 739 | |||
XM_006238538.2 | XP_006238600.1 | 5, 6 | 739 | |||
XM_008763907.1 | XP_008762129.1 | 5, 6 | 2392 | |||
XM_008763908.1 | XP_008762130.1 | 5, 6 | 2390 | |||
XM_008763909.1 | XP_008762131.1 | 5, 6 | 2387 | |||
GenBank mRNA | AF121893.1 | - | 5, 6 | 708 | ||
BC107666.1 | - | 5, 6 | 692 | 7936459 |