Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053379.3 | NP_445831.3 | 2 | 227 | ||
XM_006257382.2 | XP_006257444.1 | 2 | 352 | |||
XM_006257383.2 | XP_006257445.1 | 2 | 445 | |||
XM_006257384.2 | XP_006257446.1 | 2 | 444 | |||
XM_006257385.2 | XP_006257447.1 | 2 | 445 | |||
XM_006257386.2 | XP_006257448.1 | 2 | 445 | |||
XM_008773448.1 | XP_008771670.1 | 2 | 914 | |||
GenBank mRNA | AF155959.2 | - | 1 | 161 |