Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053350.2 | NP_445802.2 | 10, 11 | 1166 | ||
GenBank mRNA | AF121052.1 | - | 10, 11 | 1071 |