Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGEF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004706.3 | NP_004697.2 | 9 | 842 | ||
NM_198977.1 | NP_945328.1 | 8 | 743 | |||
NM_199002.1 | NP_945353.1 | 9 | 867 | |||
XM_005259386.4 | XP_005259443.1 | 9 | 1194 | |||
XM_005259390.4 | XP_005259447.1 | 9 | 1190 | |||
XM_011527468.2 | XP_011525770.1 | 9 | 1190 | |||
GenBank mRNA | BC005155.2 | - | 6 | 580 | ||
BC011726.2 | - | 8 | 718 | 4138058 | ||
BC034013.1 | - | 9 | 798 | 3451036 | ||
BC067262.1 | - | 8 | 686 | 6502165 | ||
BT007421.1 | - | 6 | 556 | |||
U64105.1 | - | 9 | 787 |