Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTDP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202504.1 | NP_001189433.1 | 13 | 2851 | ||
NM_001318511.1 | NP_001305440.1 | 12 | 3057 | |||
NM_004715.4 | NP_004706.3 | 13 | 3224 | |||
NM_048368.3 | NP_430255.2 | 12 | 3061 | |||
XM_011526261.1 | XP_011524563.1 | 11 | 2872 | |||
XM_017026078.1 | XP_016881567.1 | 11 | 2704 | |||
GenBank mRNA | AF081287.1 | - | 12 | 2772 | ||
AF154115.1 | - | 13 | 3160 | |||
AK123180.1 | - | 2 | 1242 | |||
BC015010.1 | - | 6 | 2008 | |||
BC032515.1 | - | 12 | 2711 | |||
BC047999.1 | - | 13 | 3224 | |||
BC052576.1 | - | 13 | 2816 | |||
BC063447.1 | - | 12 | 3016 | 5500032 | ||
BM512113.1 | - | 1 | 304 | |||
BX103692.1 | - | 1 | 300 |