Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tle3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053400.1 | NP_445852.1 | 2 | 831 | ||
XM_006243211.2 | XP_006243273.1 | 3 | 1024 | |||
XM_006243212.2 | XP_006243274.1 | 2 | 1200 | |||
XM_006243213.2 | XP_006243275.1 | 2 | 1199 | |||
XM_006243214.2 | XP_006243276.1 | 2 | 1200 | |||
XM_006243215.2 | XP_006243277.1 | 2 | 1199 | |||
XM_006243216.2 | XP_006243278.1 | 2 | 1199 | |||
XM_006243217.2 | XP_006243279.1 | 2 | 1199 | |||
XM_006243218.2 | XP_006243280.1 | 2 | 1198 | |||
XM_006243219.2 | XP_006243281.1 | 2 | 1199 | |||
XM_006243220.2 | XP_006243282.1 | 2 | 1199 | |||
XM_006243221.2 | XP_006243283.1 | 2 | 1199 | |||
XM_008766347.1 | XP_008764569.1 | 2 | 1199 | |||
GenBank mRNA | AF186092.1 | - | 2 | 831 | ||
BC097419.1 | - | 2 | 312 | 7454629 |