Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trpc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083115.1 | NP_001076584.1 | 1 | 82 | ||
NM_080396.1 | NP_536321.1 | 1 | 82 | |||
XM_006232356.2 | XP_006232418.1 | 2 | 406 | |||
XM_008761008.1 | XP_008759230.1 | 2 | 406 | |||
GenBank mRNA | AB008889.2 | - | 2 | 195 | ||
AF288407.1 | - | 1 | 98 | |||
AF288408.1 | - | 1 | 98 | |||
AF421363.1 | - | 1 | 82 | |||
AF421364.1 | - | 1 | 82 | |||
AF421365.1 | - | 1 | 82 | |||
AF421366.1 | - | 1 | 82 | |||
AF421367.1 | - | 1 | 82 | |||
AF421368.1 | - | 1 | 82 | |||
FQ228645.1 | - | 2 | 301 |