Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCEAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006639.1 | NP_001006640.1 | 3 | 1004 | ||
NM_001006640.1 | NP_001006641.1 | 3 | 1016 | |||
NM_004780.2 | NP_004771.2 | 3 | 1016 | |||
GenBank mRNA | BC000809.1 | - | 3 | 969 | 3452909 | |
M99701.1 | - | 3 | 992 |