Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSTO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191002.1 | NP_001177931.1 | 5 | 798 | ||
NM_001191003.1 | NP_001177932.1 | 6 | 950 | |||
NM_004832.2 | NP_004823.1 | 6 | 897 | |||
GenBank mRNA | AF212303.1 | - | 6 | 704 | ||
AK313680.1 | - | 6 | 715 | |||
BC000127.1 | - | 6 | 707 | 3351359 | ||
BX537431.1 | - | 6 | 825 | |||
CF130513.1 | - | 6 | 673 | |||
CN283947.1 | - | 5 | 601 | |||
EU794619.1 | - | 6 | 704 | |||
U90313.1 | - | 6 | 687 |