Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053425.1 | NP_445877.1 | 6, 7 | 626 | ||
GenBank mRNA | AF255305.1 | - | 6, 7 | 626 | ||
BC158586.1 | - | 6, 7 | 586 | 9099085 | ||
EU340033.1 | - | 5, 6 | 488 |