Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ran | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053439.1 | NP_445891.1 | 5 | 466 | ||
XM_006249285.2 | XP_006249347.1 | 5 | 453 | |||
GenBank mRNA | AF306457.1 | - | 5 | 466 | ||
BC059123.1 | - | 5 | 481 | 6920235 | ||
FQ220083.1 | - | 5 | 449 | |||
FQ220326.1 | - | 5 | 449 | |||
FQ220799.1 | - | 5 | 494 | |||
FQ229342.1 | - | 5 | 418 | |||
FQ229588.1 | - | 5 | 472 | |||
FQ229761.1 | - | 5 | 457 | |||
FQ231051.1 | - | 4 | 461 | |||
FQ233248.1 | - | 5 | 420 |