Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053491.2 | NP_445943.1 | 2, 3 | 230 | ||
GenBank mRNA | AJ242649.1 | - | 2, 3 | 233 | ||
BC091135.1 | - | 2, 3 | 230 | 7369357 | ||
FQ210878.1 | - | 2, 3 | 373 |