Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnga3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053495.1 | NP_445947.1 | 2, 3 | 241 | ||
XM_008767042.1 | XP_008765264.1 | 4, 5 | 765 | |||
XM_008767043.1 | XP_008765265.1 | 3, 4 | 651 | |||
GenBank mRNA | AB002801.1 | - | 2, 3 | 241 | ||
AJ272428.1 | - | 4, 5 | 528 | |||
AJ272429.1 | - | 3, 4 | 414 |