Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053501.2 | NP_445953.1 | 7 | 712 | ||
XM_006257531.2 | XP_006257593.1 | 6 | 705 | |||
XM_006257532.1 | XP_006257594.1 | 7 | 749 | |||
XM_006257534.2 | XP_006257596.1 | 7 | 1645 | |||
XM_006257535.1 | XP_006257597.1 | 7 | 567 | |||
XM_008773530.1 | XP_008771752.1 | 6 | 710 | |||
XM_008773531.1 | XP_008771753.1 | 8 | 602 | |||
GenBank mRNA | AB028879.1 | - | 6 | 509 | ||
AF300424.1 | - | 7 | 713 | |||
AJ300165.1 | - | 6 | 518 |