Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nme3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053507.1 | NP_445959.1 | 4, 5 | 393 | ||
XM_008767598.1 | XP_008765820.1 | 4, 5 | 505 | |||
GenBank mRNA | AY017337.1 | - | 4, 5 | 393 |