Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf103 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053438.2 | NP_445890.2 | 4 | 590 | ||
XM_006236653.2 | XP_006236715.1 | 4 | 1343 | |||
GenBank mRNA | AF306394.1 | - | 4 | 1314 | ||
BC100629.1 | - | 4 | 590 | 7938106 |