Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nucb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053463.1 | NP_445915.1 | 3 | 355 | ||
XM_006229163.1 | XP_006229225.1 | 4 | 472 | |||
XM_006229164.2 | XP_006229226.1 | 4 | 416 | |||
XM_006229165.1 | XP_006229227.1 | 4 | 472 | |||
GenBank mRNA | AF336828.1 | - | 3 | 355 | ||
BC062084.1 | - | 4 | 416 | |||
BC100643.1 | - | 4 | 405 | 7930904 | ||
Z36277.1 | - | 4 | 417 |