Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sema6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053471.2 | NP_445923.1 | 12 | 1573 | ||
XM_006244348.2 | XP_006244410.1 | 13 | 2042 | |||
XM_006244349.2 | XP_006244411.1 | 12 | 1395 | |||
GenBank mRNA | AB000776.1 | - | 12 | 1573 |