Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDKN2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004936.3 | NP_004927.2 | 2 | 1940 | ||
NM_078487.2 | NP_511042.1 | 2 | 2063 | |||
GenBank mRNA | AF488410.1 | - | 1 | 1223 | ||
AF488411.1 | - | 1485 | ||||
BC014469.1 | - | 2 | 1910 | 4871014 |