Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lyst | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053518.1 | NP_445970.1 | 3 | 1328 | ||
GenBank mRNA | AB020019.1 | - | 3 | 1328 |