Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053597.3 | NP_446049.1 | 2, 3 | 156 | ||
GenBank mRNA | AF184893.1 | - | 2, 3 | 150 | ||
BC061539.1 | - | 160 | 6918361 | |||
FQ209846.1 | - | 2, 3 | 156 | |||
FQ210151.1 | - | 2, 3 | 155 | |||
FQ217607.1 | - | 2, 3 | 158 | |||
FQ217624.1 | - | 2, 3 | 156 | |||
FQ218228.1 | - | 2, 3 | 157 | |||
FQ218581.1 | - | 2, 3 | 156 | |||
FQ221131.1 | - | 2, 3 | 156 | |||
FQ221294.1 | - | 2, 3 | 156 | |||
FQ221645.1 | - | 2, 3 | 155 | |||
FQ221664.1 | - | 2, 3 | 156 | |||
FQ221723.1 | - | 2, 3 | 155 | |||
FQ221727.1 | - | 2, 3 | 153 | |||
FQ221743.1 | - | 2, 3 | 156 | |||
FQ221809.1 | - | 2, 3 | 156 | |||
FQ221911.1 | - | 2, 3 | 156 | |||
FQ221975.1 | - | 2, 3 | 157 | |||
FQ222049.1 | - | 2, 3 | 156 | |||
FQ222054.1 | - | 2, 3 | 157 | |||
FQ222270.1 | - | 2, 3 | 157 | |||
FQ222348.1 | - | 3, 4 | 273 | |||
FQ222848.1 | - | 2, 3 | 156 | |||
FQ222865.1 | - | 2, 3 | 156 | |||
FQ222911.1 | - | 2, 3 | 156 | |||
FQ223072.1 | - | 2, 3 | 156 | |||
FQ223291.1 | - | 2, 3 | 155 | |||
FQ223549.1 | - | 2, 3 | 156 | |||
FQ223731.1 | - | 2, 3 | 157 | |||
FQ223862.1 | - | 2, 3 | 156 | |||
FQ224473.1 | - | 2, 3 | 156 | |||
FQ228507.1 | - | 2, 3 | 157 | |||
FQ228599.1 | - | 2, 3 | 156 | |||
FQ228823.1 | - | 2, 3 | 156 | |||
FQ229526.1 | - | 2, 3 | 156 |
Gene Symbol: LOC100362987 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002730094.4 | XP_002730140.1 | 1 | 123 |
Gene Symbol: LOC100364561 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006249411.2 | XP_006249473.1 | 1 | 259 |
Gene Symbol: LOC100911728 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751271.3 | XP_003751319.1 | 1 | 205 |
Gene Symbol: LOC102554672 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008763965.1 | XP_008762187.1 | 2 | 195 |
Gene Symbol: Slfn14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY539914.1 | - | 123 |