Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nupr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053611.2 | NP_446063.1 | 2 | 224 | ||
GenBank mRNA | AA685246.1 | - | 2 | 224 | ||
AF014503.1 | - | 2 | 192 |
Gene Symbol: NEWGENE_69363 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008759924.1 | XP_008758146.1 | 2 | 275 |