Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rtn4r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053613.1 | NP_446065.1 | 2 | 1255 | ||
XM_008768839.1 | XP_008767061.1 | 2 | 1788 | |||
GenBank mRNA | AF462390.1 | - | 2 | 1255 | ||
AY028438.1 | - | 2 | 1255 |