Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005068.2 | NP_005059.2 | 11 | 3983 | ||
XM_005267100.2 | XP_005267157.1 | 12 | 4559 | |||
XM_017011197.1 | XP_016866686.1 | 12 | 4291 |