Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnj13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053608.2 | NP_446060.1 | 3 | 755 | ||
XM_008767301.1 | XP_008765523.1 | 2 | 2090 | |||
GenBank mRNA | AJ006129.1 | - | 2 | 638 |