Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Magi2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053621.1 | NP_446073.1 | 1, 2 | 462 | ||
GenBank mRNA | AF034863.1 | - | 1, 2 | 462 |