Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sfrp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053544.1 | NP_445996.1 | 4, 5 | 893 | ||
GenBank mRNA | AF012891.1 | - | 4, 5 | 893 | ||
AF140346.1 | - | 4, 5 | 1044 | |||
AF220608.1 | - | 1141 | ||||
FQ220237.1 | - | 1, 2 | 202 | |||
FQ229078.1 | - | 1, 2 | 215 | |||
FQ229224.1 | - | 3, 4 | 324 |
Set Membership: |
G-protein coupled receptor |