Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clic5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053603.2 | NP_446055.1 | 5 | 741 | ||
GenBank mRNA | AF323174.1 | - | 5 | 749 |