Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccnl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053662.3 | NP_446114.1 | 424 | |||
GenBank mRNA | AF030091.3 | - | 424 | |||
BC085686.1 | - | 405 | ||||
BC128699.1 | - | 371 | ||||
BC161817.1 | - | 370 | 9022416 | |||
CB608234.1 | - | 431 | ||||
FQ232436.1 | - | 388 |
Gene Symbol: LOC100909712 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749295.3 | XP_003749343.1 | 2 | 733 |