Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Becn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034117.1 | NP_001029289.1 | 2, 3 | 327 | ||
NM_053739.2 | NP_446191.1 | 2, 3 | 349 | |||
GenBank mRNA | AY033824.1 | - | 1, 2 | 125 | ||
BC074011.1 | - | 2, 3 | 214 | 7100185 | ||
CB788828.1 | - | 2, 3 | 364 | |||
CK600131.1 | - | 2, 3 | 327 |