Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dlk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053744.1 | NP_446196.1 | 378 | |||
XM_008764917.1 | XP_008763139.1 | 434 | ||||
GenBank mRNA | BC167752.1 | - | 398 | 8365650 | ||
D84336.1 | - | 378 | ||||
U25680.1 | - | 243 |